DNA transfection reagent, EcoTransfect™
EcoTransfect™ transfection reagent is an economical lipofection reagent dedicated to the transfection of popular cell lines. It is the perfect solution for quickly analysing the biological activity of nucleic acids, to perform routine transfection assays at low cost and to accomplish high throughput screening.
- Economical
- Good efficiency
- Serum compatible
- Simple, ready to use and rapid
Store at +4 °C.
: Shipped at room temperature.
- DNA:Yes
- Oligos:Yes