Order Entry
Germany
ContactUsLinkComponent
 
Sodium chloride ≥99.0% ACS, Ultrapure
 
Sodium chloride ≥99.0% ACS, Ultrapure
 

 

  • Assay Chlorate and Nitrate (NO₃):
    ≤0.003%
  • Assay:
    ≥99.0%
  • Residual Halide Bromide (Br):
    ≤0.01%
  • Residual Halide Iodide (I):
    ≤0.002%
  • Impurity Content Phosphate (PO₄):
    ≤5 ppm
  • Impurity Content Sulphate (SO₄):
    ≤0.004%
  • Insoluble Material:
    ≤0.005%
  • pH (5%, H₂O, 25 °C):
    5.0 - 9.0
  • Barium (Ba):
    Passes Test
  • Calcium (Ca):
    ≤0.002%
  • Heavy Metal (Pb):
    ≤5 ppm
  • Iron (Fe):
    ≤2 ppm
  • Magnesium (Mg):
    ≤0.001%
  • Potassium (K):
    ≤0.005%