Guanosine-3',5'-cyclicmonophosphoric acid sodium salt ≥98% (by HPLC), Millipore®
Catalog # 370656-50
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Plastic ampoule
- Pk:50 mg
- Seq:2370
Specifications
Specification Test Results
- Purity (HPLC):≥98%
Specification Test Results
About this item
:
:367.19 g/mol
Storage Temperature:Freezer
CAS Number:40732-48-7
Formula:C₁₀H₁₁N₅NaO₇P
MDL Number:MFCD00038424