Pellet presses
Catalog # PARR2814
Supplier: PARR
Specifications
- Description:Pellet press, Ø: 1/8"
- Seq:2
- Pk:1
Specifications
About this item
Pellet presses used for compressing powdered samples into pellet or tablet form for oxygen bomb calorimetry.
- Pellets produced in cylindrical shape with flat ends
Suitable for pressing powdered material into briquette form.