Polyethylene glycol diacrylate (PEGDA) ≥92.0% Mn 250 inhibited, Sigma-Aldrich®
: Merck
:26570-48-9
:
:CH₂=COC(OCH₂CH₂)nOCOC=CH₂
:MFCD00081876
:1.11 g/ml (25 °C)
:M-Clarity™ quality level = MQ100
:Stabilised with 4-methoxyphenol (MEHQ) 100 ppm
:GHS05
:GHS07
:Refrigerator
- Refractive Index:n20/D 1.463
- Ω-end:Acrylate
- α-end:Acrylate
- Water (by Karl Fischer):≤1.00%
- Acid value:≤1.00 (mg KOH/g)
- Color test:≤200 APHA
- Inhibitor (MEHQ):80 - 120 ppm
- Purity:≥92.0%
- Proton NMR Spectra:Conforms to Structure
- Polymer Architecture (Shape):Linear
- Polymer Architecture (Functionality):Homobifunctional
- Reaction Suitability (Reagent Type):Cross-linking Reagent
- Reaction Suitability (Reaction Type):Polymerisation Reactions