Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DL-Glyceraldehyde
DL-Glyceraldehyde ≥90% (by GC), Sigma-Aldrich®
DL-Glyceraldehyde ≥90% (by GC), Sigma-Aldrich®
DL-Glyceraldehyde ≥90% (by GC), Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
DL-Glyceraldehyde ≥90% (by GC), Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
56-82-6
:
Formula:
C₃H₆O₃
MDL Number:
MFCD00064379
:
90.078 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
200-290-0