
Folin-Ciocalteu's reagent for analysis of phenols
Catalog # VWRS31360.320
Supplier: VWR CUSTOM MADE CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
Density:1.22 g/cm³ (20 °C)
UN:3264
ADR:8,III
:GHS07
MDL Number:MFCD00132625