
Poly(allylamine hydrochloride), M.W. 50000, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71550-12-4
:
Formula:[CH₂CH(CH₂NH₂·HCl)]n
MDL Number:MFCD00084396
:93.5600 g/mol
:M-Clarity™ quality level = MQ100
EINECS:415-050-2
:GHS07
Specification Test Results
- 1H NMR Spectrum:Conforms to Structure
- Molecular Weight:40000 - 60000
- Residual Monomer:≤10%
Specification Test Results
Frequently Bought Together