Order Entry
Germany
Orders LinkContactUsLinkComponent
Buffer solution pH 10

Buffer solution pH 10

Supplier:   Bernd Kraft
Buffer solution pH 10
Supplier:  Bernd Kraft
Certificates
Buffer solution pH 10

6 Options Available Below

Product Details & Documents

This solution is a combination of 53,5 g NH₄Cl, 350 ml NH₃ solution 25% and Cu-EDTA/l used for complexometric determination of zinc.

Product Family Options

Product Information

  • pH value
    pH buffer type
    Pk
    Availability
    Price
  • KRAF03596.2000
    Buffer solution
    Buffer solution
    10
    Ammonia
    0,5 L
    Log in for availability
    Log in for availability
    1 * 0,5 L
    1 * 0,5 L

  • KRAF03596.3000
    Buffer solution
    Buffer solution
    10
    Ammonia
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 66,80 €
    66,80 €
    1 * 1 L
       66,80 €

  • KRAF03596.5020
    Buffer solution
    Buffer solution
    10
    Ammonia
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L

  • KRAF03596.5300
    Buffer solution
    Buffer solution
    10
    Ammonia
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 190,00 €
    190,00 €
    1 * 5 L
       190,00 €

  • KRAF03596.6000
    Buffer solution
    Buffer solution
    10
    Ammonia
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L - 310,00 €
    310,00 €
    1 * 10 L
       310,00 €

  • KRAF03596.7020
    Buffer solution
    Buffer solution
    10
    Ammonia
    20 L
    Log in for availability
    Log in for availability
    1 * 20 L
    1 * 20 L