Order Entry
Germany
Orders LinkContactUsLinkComponent
Autosampler vials

Autosampler vials

Crimp top vials, with wide opening, 0,30 ml, 12×13 mm, Glass, Clear, No
Catalog # MICS9532C-0-WCV
Supplier:   MICROSOLV
Autosampler vials
Catalog # MICS9532C-0-WCV
Supplier:  MICROSOLV
Autosampler vials
Autosampler vials

Crimp top vials, with wide opening, 0,30 ml, 12×13 mm, Glass, Clear, No

Price
391,00 €
Log in for availability
1 * 100 ST
Product Details & Documents

These vials with serum finish are made from Type 1, first hydrolytic N51A grade borosilicate glass.

  • Used in gas chromatography
  • Used for storage of samples

Standard crimp top style vials for HPLC or GC autosamplers.

Specifications
  • Cap included:
    No
  • Ø×H:
    12×13 mm
  • Capacity:
    0,30 ml
  • Colour:
    Clear
  • Description:
    Crimp top vials, with wide opening
  • Material:
    Glass
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.