Order Entry
Germany
ContactUsLinkComponent
 
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: 8G7G3/1]
 
Anti-NKX2-1 Mouse Monoclonal Antibody [clone: 8G7G3/1]

 

  • Catalog No:
  • ILM4311-C1
  • ILM4311-C05
  • ILM4311-C01
  • Antigen symbol:
  • NKX2-1
  • NKX2-1
  • NKX2-1
  • Antigen name:
  • NK2 homeobox 1
  • NK2 homeobox 1
  • NK2 homeobox 1
  • Antigen synonyms:
  • TTF-1|BCH|NKX2A|TTF1|TITF1
  • TTF-1|BCH|NKX2A|TTF1|TITF1
  • TTF-1|BCH|NKX2A|TTF1|TITF1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 8G7G3/1
  • 8G7G3/1
  • 8G7G3/1
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 7080
  • 7080
  • 7080
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

 

Frequently Bought Together