Anti-TIA1 Mouse Monoclonal Antibody [clone: 2G9A10F5]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: TIA1
Clonality: Monoclonal
Clone: 2G9A10F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ILM25523-C1
- ILM25523-C05
- ILM25523-C01
- Antigen symbol:
- TIA1
- TIA1
- TIA1
- Antigen name:
- TIA1 cytotoxic granule associated RNA binding protein
- TIA1 cytotoxic granule associated RNA binding protein
- TIA1 cytotoxic granule associated RNA binding protein
- Antigen synonyms:
- Granule membrane protein (GMP-17)
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- 2G9A10F5
- 2G9A10F5
- 2G9A10F5
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 7072
- 7072
- 7072
- Isotype:
- IgG1
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- Human bone marrow malignant cells from a non-B, non-T acute leukemia
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 17 kDa
- Purification:
- Ion exchange or affinity chromatography
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together