Order Entry
Germany
ContactUsLinkComponent
 
Anti-TAG-72 Mouse Monoclonal Antibody [clone: B72.3]
 
Anti-TAG-72 Mouse Monoclonal Antibody [clone: B72.3]

 

  • Catalog No:
  • ILM7231-C1
  • ILM7231-C05
  • ILM7231-C01
  • Antigen symbol:
  • TAG-72
  • TAG-72
  • TAG-72
  • Antigen name:
  • Tumor associated glycoprotein 72
  • Tumor associated glycoprotein 72
  • Tumor associated glycoprotein 72
  • Antigen synonyms:
  • CA72.4
  • CA72.4
  • CA72.4
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • B72.3
  • B72.3
  • B72.3
  • Reactivity:
  • Hamster, Human, Bovine, Dog, Rat
  • Hamster, Human, Bovine, Dog, Rat
  • Hamster, Human, Bovine, Dog, Rat
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Molecular weight:
  • 220 kDa
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

 

Frequently Bought Together