Order Entry
Germany
ContactUsLinkComponent
 
Anti-SOX11 Mouse Monoclonal Antibody [clone: ILQ-158]
 
Anti-SOX11 Mouse Monoclonal Antibody [clone: ILQ-158]

 

  • Catalog No:
  • ILM3823-C1
  • ILM3823-C05
  • ILM3823-C01
  • Antigen symbol:
  • SOX11
  • SOX11
  • SOX11
  • Antigen name:
  • SRY-box 11
  • SRY-box 11
  • SRY-box 11
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • ILQ-158
  • ILQ-158
  • ILQ-158
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 6664
  • 6664
  • 6664
  • Isotype:
  • IgG1
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

 

Frequently Bought Together