Anti-S100B Mouse Polyclonal Antibody [clone: 4C4.9]
Type: Primary
Antigen: S100B
Clonality: Polyclonal
Clone: 4C4.9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat, Cow
- Catalog No:
- ILM3303-C1
- ILM3303-C05
- ILM3303-C01
- Antigen symbol:
- S100B
- S100B
- S100B
- Antigen name:
- S100 calcium binding protein B
- S100 calcium binding protein B
- S100 calcium binding protein B
- Antigen synonyms:
- S100beta
- S100beta
- S100beta
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- 4C4.9
- 4C4.9
- 4C4.9
- Reactivity:
- Human, Bovine, Rat, Mouse
- Human, Bovine, Rat, Mouse
- Human, Bovine, Rat, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 6285
- 6285
- 6285
- Isotype:
- IgG2a kappa
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Frequently Bought Together