Order Entry
Germany
ContactUsLinkComponent
Anti-TP63 Mouse Monoclonal Antibody [clone: 4A4]
Anti-TP63 Mouse Monoclonal Antibody [clone: 4A4]
Supplier:  IMMUNOLOGIC
Certificates
Anti-TP63 Mouse Monoclonal Antibody [clone: 4A4]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM8651-C01
  • ILM8651-C05
  • ILM8651-C1
  • Antigen symbol:
  • TP63
  • TP63
  • TP63
  • Antigen name:
  • Tumor protein p63
  • Tumor protein p63
  • Tumor protein p63
  • Antigen synonyms:
  • p51|p73L|OFC8|SHFM4|p63|EEC3|KET|p53CP|p73H|TP73L|NBP|TP53CP|TP53L
  • p51|p73L|OFC8|SHFM4|p63|EEC3|KET|p53CP|p73H|TP73L|NBP|TP53CP|TP53L
  • p51|p73L|OFC8|SHFM4|p63|EEC3|KET|p53CP|p73H|TP73L|NBP|TP53CP|TP53L
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 4A4
  • 4A4
  • 4A4
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 8626
  • 8626
  • 8626
  • Isotype:
  • IgG kappa
  • IgG kappa
  • IgG kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

Specifications

Related Information

Frequently Bought Together