Order Entry
Germany
ContactUsLinkComponent
Anti-NF Mouse Monoclonal Antibody [clone: 2F11]
Anti-NF Mouse Monoclonal Antibody [clone: 2F11]
Supplier:  IMMUNOLOGIC
Certificates
Anti-NF Mouse Monoclonal Antibody [clone: 2F11]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM1503-C1
  • ILM1503-C05
  • ILM1503-C01
  • Antigen symbol:
  • NF
  • NF
  • NF
  • Antigen name:
  • Neurofilament
  • Neurofilament
  • Neurofilament
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2F11
  • 2F11
  • 2F11
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C

Specifications

Related Information

Frequently Bought Together