Anti-MYOG Mouse Monoclonal Antibody [clone: F5D]
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: F5D
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Dog, Cat, Pig
- Catalog No:
- ILM2963-C1
- ILM2963-C05
- ILM2963-C01
- Antigen symbol:
- MYOG
- MYOG
- MYOG
- Antigen name:
- Myogenin
- Myogenin
- Myogenin
- Antigen synonyms:
- MYF4|bHLHc3
- MYF4|bHLHc3
- MYF4|bHLHc3
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- F5D
- F5D
- F5D
- Reactivity:
- Pig, Human, Dog, Rat, Mouse, Cat
- Pig, Human, Dog, Rat, Mouse, Cat
- Pig, Human, Dog, Rat, Mouse, Cat
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4656
- 4656
- 4656
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Frequently Bought Together