Anti-MSH2 Mouse Monoclonal Antibody [clone: G219-11229]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: MSH2
Clonality: Monoclonal
Clone: G219-11229
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- ILM8734-C01
- ILM8734-C05
- ILM8734-C1
- Antigen symbol:
- MSH2
- MSH2
- MSH2
- Antigen name:
- MutS homolog 2
- MutS homolog 2
- MutS homolog 2
- Antigen synonyms:
- COCA1|HNPCC|HNPCC1
- COCA1|HNPCC|HNPCC1
- COCA1|HNPCC|HNPCC1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- G219-11229
- G219-11229
- G219-11229
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4463
- 4463
- 4463
- Isotype:
- IgG1
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together