Anti-MLANA Mouse Monoclonal Antibody [clone: A103]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: MLANA
Clonality: Monoclonal
Clone: A103
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Dog
Specifications
- Catalog No:
- ILM022111-C1
- ILM022111-C05
- ILM022111-C01
- Antigen symbol:
- MLANA
- MLANA
- MLANA
- Antigen name:
- Melan-A
- Melan-A
- Melan-A
- Antigen synonyms:
- MART1
- MART1
- MART1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- A103
- A103
- A103
- Reactivity:
- Human, Dog, Rat, Mouse
- Human, Dog, Rat, Mouse
- Human, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 2315
- 2315
- 2315
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Immunogen:
- Recombinant hMART-1 protein
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 20-22 kDa (doublet)
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
- Tested applications:
- Immunohistochemical Staining of Formalin-Fixed Tissue Sections
Specifications
Frequently Bought Together