Anti-SCGB2A2 Mouse Monoclonal Antibody [clone: 304-1A5]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: SCGB2A2
Clonality: Monoclonal
Clone: 304-1A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rabbit
Specifications
- Catalog No:
- ILM2804-C1
- ILM2804-C05
- ILM2804-C01
- Antigen symbol:
- SCGB2A2
- SCGB2A2
- SCGB2A2
- Antigen name:
- Secretoglobin family 2A member 2
- Secretoglobin family 2A member 2
- Secretoglobin family 2A member 2
- Antigen synonyms:
- MGC71974|MGB1|UGB2
- MGC71974|MGB1|UGB2
- MGC71974|MGB1|UGB2
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- 304-1A5
- 304-1A5
- 304-1A5
- Reactivity:
- Rabbit, Monkey, Human, Mouse
- Rabbit, Monkey, Human, Mouse
- Rabbit, Monkey, Human, Mouse
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4250
- 4250
- 4250
- Isotype:
- IgG1
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together