Order Entry
Germany
ContactUsLinkComponent
Anti-IGKC Mouse Monoclonal Antibody [clone: L1C1]
Anti-IGKC Mouse Monoclonal Antibody [clone: L1C1]
Supplier:  IMMUNOLOGIC
Certificates
Anti-IGKC Mouse Monoclonal Antibody [clone: L1C1]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM5103-C1
  • ILM5103-C05
  • ILM5103-C01
  • Antigen symbol:
  • IGKC
  • IGKC
  • IGKC
  • Antigen name:
  • Immunoglobulin kappa constant
  • Immunoglobulin kappa constant
  • Immunoglobulin kappa constant
  • Antigen synonyms:
  • IGKC|HCAK1
  • IGKC|HCAK1
  • IGKC|HCAK1
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • L1C1
  • L1C1
  • L1C1
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 3514
  • 3514
  • 3514
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Human B-Lymphoma cells
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Molecular weight:
  • 22,5 kDa
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C
  • Tested applications:
  • Immunohistochemical Staining of Formalin-Fixed Tissue Sections

Specifications

Related Information

Frequently Bought Together