Anti-HSA Mouse Monoclonal Antibody [clone: OCH 1E5]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: HSA
Clonality: Monoclonal
Clone: OCH 1E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog
Specifications
- Catalog No:
- ILM1333-C1
- ILM1333-C05
- ILM1333-C01
- Antigen symbol:
- HSA
- HSA
- HSA
- Antigen name:
- Hepatocyte specific antigen
- Hepatocyte specific antigen
- Hepatocyte specific antigen
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- OCH 1E5
- OCH 1E5
- OCH 1E5
- Reactivity:
- Human, Dog
- Human, Dog
- Human, Dog
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- IgG1
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together