Anti-MUC1 Mouse Monoclonal Antibody [clone: E29]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: E29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Specifications
- Catalog No:
- ILM1233-C1
- ILM1233-C05
- ILM1233-C01
- Antigen symbol:
- MUC1
- MUC1
- MUC1
- Antigen name:
- Mucin 1/cell surface associated
- Mucin 1/cell surface associated
- Mucin 1/cell surface associated
- Antigen synonyms:
- PUM|MCKD|ADMCKD1|MCD|CD227|PEM|ADMCKD|MCKD1
- PUM|MCKD|ADMCKD1|MCD|CD227|PEM|ADMCKD|MCKD1
- PUM|MCKD|ADMCKD1|MCD|CD227|PEM|ADMCKD|MCKD1
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- E29
- E29
- E29
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 4582
- 4582
- 4582
- Isotype:
- IgG2a kappa
- IgG2a kappa
- IgG2a kappa
- ImmunoChemistry:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Form:
- Concentrated
- Concentrated
- Concentrated
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together