Anti-KRT5 Mouse Monoclonal Antibody [clone: 10C11E6]
Supplier: IMMUNOLOGIC
Certificates
About this item
Type: Primary
Antigen: KRT5
Clonality: Monoclonal
Clone: 10C11E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ILM6831-C1
- ILM6831-C05
- ILM6831-C01
- Antigen symbol:
- KRT5
- KRT5
- KRT5
- Antigen name:
- Keratin 5
- Keratin 5
- Keratin 5
- Antigen synonyms:
- EBS2|KRT5A
- EBS2|KRT5A
- EBS2|KRT5A
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Unconjugated
- Clone:
- 10C11E6
- 10C11E6
- 10C11E6
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Gene ID:
- 3852
- 3852
- 3852
- Isotype:
- IgG
- IgG
- IgG
- Western blot:
- Yes
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Yes
- Epitope:
- 316 to 590
- Amino acid number:
- 316 to 590
- Form:
- Concentrated
- Concentrated
- Concentrated
- Molecular weight:
- 62,4 kDa
- Storage temperature:
- 2°C - 8°C
- 2°C - 8°C
- 2°C - 8°C
Specifications
Frequently Bought Together