Order Entry
Germany
ContactUsLinkComponent
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Supplier:  IMMUNOLOGIC
Certificates
Anti-PTPRC Mouse Monoclonal Antibody [clone: UCHL-1]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM07211-C1
  • ILM07211-C05
  • ILM07211-C01
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein tyrosine phosphatase/receptor type C
  • Protein tyrosine phosphatase/receptor type C
  • Protein tyrosine phosphatase/receptor type C
  • Antigen synonyms:
  • CD45|GP180|T200|LCA
  • CD45|GP180|T200|LCA
  • CD45|GP180|T200|LCA
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • UCHL-1
  • UCHL-1
  • UCHL-1
  • Reactivity:
  • Common marmoset, Chimpanzee, Human
  • Common marmoset, Chimpanzee, Human
  • Common marmoset, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 5788
  • 5788
  • 5788
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • Cultured human T-cells from an IL-2-dependent T-cell line (CA1)
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Molecular weight:
  • 180-185 kDa
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C
  • Tested applications:
  • Immunohistochemical Staining of Formalin-Fixed Tissue Sections

Specifications

Related Information

Frequently Bought Together