Order Entry
Germany
ContactUsLinkComponent
Anti-Actin Mouse Monoclonal Antibody [clone: HHF35]
Anti-Actin Mouse Monoclonal Antibody [clone: HHF35]
Supplier:  IMMUNOLOGIC
Certificates
Anti-Actin Mouse Monoclonal Antibody [clone: HHF35]
Supplier:  IMMUNOLOGIC

Specifications

  • Catalog No:
  • ILM1463-C1
  • ILM1463-C05
  • ILM1463-C01
  • Antigen symbol:
  • ACTA1/ACTA2/ACTG2
  • ACTA1/ACTA2/ACTG2
  • ACTA1/ACTA2/ACTG2
  • Antigen name:
  • Actin
  • Actin
  • Actin
  • Antigen synonyms:
  • Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
  • Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
  • Skeletal muscle alpha actin-1|ACTC1|ACTA2|alpha skeletal muscle|ACTA1|Alpha-actin-1|Actin|Alpha-2 actin|ACTA|Cardiac muscle alpha actin-1|ACTSA
  • Antibody type:
  • Primary
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Unconjugated
  • Clone:
  • HHF35
  • HHF35
  • HHF35
  • Reactivity:
  • Rabbit, Chicken, Human, Dog, Rat, Mouse, Cat
  • Rabbit, Chicken, Human, Dog, Rat, Mouse, Cat
  • Rabbit, Chicken, Human, Dog, Rat, Mouse, Cat
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Gene ID:
  • 58, 59, 70
  • 58, 59, 70
  • 58, 59, 70
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Yes
  • Immunogen:
  • SDS extract of human myocardium
  • Form:
  • Concentrated
  • Concentrated
  • Concentrated
  • Storage temperature:
  • 2°C - 8°C
  • 2°C - 8°C
  • 2°C - 8°C
  • Tested applications:
  • Immunohistochemical Staining of Formalin-Fixed Tissue Sections

Specifications

Related Information

Frequently Bought Together