


1,2-Dichloroethane ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6726-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):min. 99.8 %
- Residue after Evaporation:max. 0.0005 %
- Titrable Acid (meq/g):max. 0.0005
- Water (KF):max. 0.01 %
- at 240 nm:max. 0.3
- at 250 nm:max. 0.07
- at 270 nm:max. 0.009
Specification Test Results
About this item
:
:98.96 g/mol
Boiling Pt:84 °C
Flash Pt:13 °C
Storage Temperature:Ambient
UN:1486
ADR:3(6.1),II
:GHS02
:GHS07
:GHS08
CAS Number:107-06-2
Formula:ClH₂CCH₂Cl
MDL Number:MFCD00000963