Order Entry
Germany
ContactUsLinkComponent
Chloroform ≥99.8% stabilised, ChromAR® for HPLC, Macron Fine Chemicals™
Chloroform ≥99.8% stabilised, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 4443-25
CAS Number:  
undefined
Chloroform ≥99.8% stabilised, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 4443-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Assay ( GC, corrected for water):
    min. 99.8 %
  • Residue after Evaporation:
    max. 0.0005 %
  • Stabiliser (Amylene):
    ~ 50 ppm
  • Titrable Acid (meq/g):
    max. 0.0005
  • Water:
    max. 0.01 %
  • at 450 nm:
    max. 1 ppb
  • at 250 nm:
    max. 0.30
  • at 260 nm:
    max. 0.07
  • at 270 nm:
    max. 0.01
  • at 280 nm:
    max. 0.004

Specification Test Results

About this item

 :
 :119.38 g/mol
Boiling Pt:62 °C
Density:1.48
ADR:6.1,III
 :Stabilised with amylene ~ 50 ppm
EINECS:200-663-8
UN:1888
 :GHS07
 :GHS08
CAS Number:67-66-3
Formula:CHCl₃
MDL Number:MFCD00000826