

2-Propanol ≥99.8%, ChromAR® for HPLC, Macron Fine Chemicals™
3043-23
:
- Pack type:NOWPAK® package, returnable
- Pk:20 L
- Seq:5302
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Assay (by GC):min. 99.8 %
- Assay (GC):min. 99.8 %
- Residue after Evaporation:max. 0.0005 %
- Residue on Evaporation:max. 0.0005 %
- Titrable Acid (meq/g):max. 0.0005
- Titrable Base (meq/g):max. 0.0005
- Water (KF):max. 0.05 %
- at 210 nm:max. 0.4
- at 220 nm:max. 0.2
- at 230 nm:max. 0.1
- at 240 nm:max. 0.05
- at 250 nm:max. 0.02
- UV Cut-off:max. 203 nm
:
:60.1 g/mol
:13,05228
:82 °C (1013 hPa)
:–89 °C
:0.786 g/cm³ (20 °C)
:12 °C
:Ambient
:200-661-7
:1219
:3,II
:01-2119457558-25
:GHS02
:GHS07
:67-63-0
:(CH₃)₂CHOH
:MFCD00011674