Synthetic [D-Ala²]-leucine enkephalin
SIALE5008-5MG
: Merck
:
- Pack type:Serum bottle
- Pk:5 mg
- CAS No.:64963-01-5
- Protein/peptide type:Synthetic
- Storage conditions:−20 °C
- Form:Powder
- Protein synonyms:Tyr-D-Ala-Gly-Phe-Leu
- Protein/peptide name:[D-Ala2]-leucine enkephalin
- Purity:>97%
- Molecular weight:569,65 g/mol
- Sequence:Tyr-Ala-Gly-Phe-Leu
- Colour:White
- Molecular formula:C₂₉H₃₉N₅O₇
- Purification:HPLC
Potent, long-acting Leu-enkephalin analog that is resistant to enzymatic degradation.
- Amino acid sequence: Tyr-Ala-Gly-Phe-Leu
The enkephalin amid peptide [D-Ala²]-leucine enkephalin ((DADLE), a delta opioid agonist, is a degradation resistant long-acting Leu-enkephalin analog used to study the signaling pathway of delta opioid receptors.