Safety knife, SECUMAX 145
Catalog # MARO145001.12
Supplier: MARTOR ARGENTAX
Specifications
- Description:Safety knife, SECUMAX 145
- Seq:02
- Total length:118 mm
- Pk:10
Specifications
About this item
The SECUMAX 145 is a compact safety knife for routine cutting tasks. The two cutting edges are safely concealed to protect fingers.
- Blade made of glass fibre reinforced plastic
- Two in one blade head for cutting and piercing
The disposable cutting tool easily copes with single-layer cardboard, film and packing tape, for example.