Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(R)-2-Acetoxy-2-phenylacetic acid
(R)-2-Acetoxy-2-phenylacetic acid 98% (by HPLC)
(R)-2-Acetoxy-2-phenylacetic acid 98% (by HPLC)
:
Thermo Fisher Scientific
(R)-2-Acetoxy-2-phenylacetic acid 98% (by HPLC)
:
Thermo Fisher Scientific
:
51019-43-3
:
:
C₁₀H₁₀O₄
:
MFCD00004249
:
97…99 °C
:
194.19 g/mol
:
GHS07
:
Ambient
Assay (HPLC):
97.5%
msds_ALFAH56492
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years