VWR® PerfectBlue, Tank Blotters
Supplier: VWR Collection
Certificates
About this item
PerfectBlue S and M tank electroblotters are designed to rapidly transfer nucleic acid or protein fragments from up to four polyacrylamide gels at a time to nitrocellulose, nylon or PVDF membranes. The colour coded cassettes allow for easy assembly of transfer sandwich and error-free transfer. The large buffer capacity and integral cooling allow extended transfer times and high voltage/high current operation for the transfer of high molecular weight proteins.
- Sturdy buffer chamber for up to four colour coded blotting cartridges, with sufficient volume for magnetic stirrer to ensure uniform temperature and pH conditions
- Highly durable, high grade steel and platinum coated titanium electrodes give uniform electrical field over the entire transfer surface; gold plated plugs and corrosion proof electrodes for long durability and reliable electrical contact
- Water circulation system for buffer cooling at the base of the tank with effective heat exchanger made of ceramic aluminium and increased exchange surface area to aid heat transfer
- Safety cover includes securely attached cables to enhance user safety
Specifications
- Catalog No:
- 700-1230
- 700-1228
- Model:
- PerfectBlue M
- PerfectBlue S
- Buffer volume:
- ~4500 ml
- ~1300 ml
- Active transfer area:
- 360 cm³
- 80,75 cm³
- Transfer time:
- 120 - 360 min
- 120 - 360 min
- Unit W×D×H:
- 290×140×350 mm
- 185×145×155 mm