3-Methoxythiophene
1
Catalog # CHMPFL13627.POR
Supplier: CHEMPUR
Jump to:
Product Details & Documents
- CAS Number:17573-92-1
- Formula:C₅H₆OS
- MDL Number:MFCD00043894
- Molecular Weight:114.17 g/mol
- Boiling Pt:95…97 °C (80 mmHg)
- Melting Pt:–7 °C
- Density:1.385 g/cm³ (–173.15 °C)
- UN:3271
- ADR:3,III
Specifications
Specifications
- Pk:1
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



