

Hexamethylenetetramine
Catalog # MOLEM38908959
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
Merck Index:13,05994
Melting Pt:280 °C
Density:1.33 g/cm³ (20 °C)
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:202-905-8
UN:1328
ADR:4.1,III
:140.19 g/mol
:GHS02
:GHS07
CAS Number:100-97-0
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895