

Hexamethylenetetramine analytical reagent
Supplier: Bernd Kraft
Certificates
About this item
CAS Number:100-97-0
:
Formula:C₆H₁₂N₄
MDL Number:MFCD00006895
Merck Index:13,05994
Melting Pt:280 °C
Density:1.33 g/cm³ (20 °C)
Flash Pt:250 °C
Storage Temperature:Ambient
EINECS:202-905-8
ADR:4.1,III
:140.19 g/mol
UN:1328
:GHS02
:GHS07