Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methenamine
Hexamethylenetetramine, Sigma-Aldrich®
Hexamethylenetetramine, Sigma-Aldrich®
:
Merck
Hexamethylenetetramine, Sigma-Aldrich®
:
Merck
:
100-97-0
:
:
C₆H₁₂N₄
:
MFCD00006895
:
13,05994
:
280 °C
:
1.33 g/cm³ (20 °C)
:
250 °C
:
202-905-8
:
1328
:
4.1,III
:
M-Clarity™ quality level = MQ100
:
140.19 g/mol
:
GHS02
:
GHS07
:
Ambient