Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,1,1,3,5,5,5-Heptamethyltrisiloxane
1,1,1,3,5,5,5-Heptamethyltrisiloxane, Sigma-Aldrich®
1,1,1,3,5,5,5-Heptamethyltrisiloxane, Sigma-Aldrich®
1,1,1,3,5,5,5-Heptamethyltrisiloxane, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,1,1,3,5,5,5-Heptamethyltrisiloxane, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
1873-88-7
:
Formula:
C₇H₂₂O₂Si₃
MDL Number:
MFCD00048000
:
222.51 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
217-496-1