
Hexamethyldisiloxane, 0.65 cSt (25°C), Sigma-Aldrich®
SIAL326739-100G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
:
:162.38 g/mol
:101 °C (1013 hPa)
:–59 °C
:0.764 g/cm³ (20 °C)
:0.6 °C (closed cup)
:Ambient
:203-492-7
:1993
:3,II
:M-Clarity™ quality level = MQ100
:01-2119496108-31
:GHS02
:107-46-0
:C₆H₁₈OSi₂
:MFCD00008265