

Hexamethyldisiloxane, 0.65 cSt (25°C) for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:107-46-0
:
Formula:C₆H₁₈OSi₂
MDL Number:MFCD00008265
:162.38 g/mol
Boiling Pt:101 °C (1013 hPa)
Melting Pt:–59 °C
Density:0.764 g/cm³ (20 °C)
Flash Pt:0.6 °C (closed cup)
Storage Temperature:Ambient
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:203-492-7
REACH:01-2119496108-31
:GHS02
:GHS09
Specification Test Results
- Melting Point:-68 °C
- Molar Mass:162.38 g/mol
- Boiling Point:100 °C (1013 hPa)
- Vapor Pressure:20 hPa (20 °C)
- Flash Point:4 °C
- Refractive Index:1.3771 (20 °C, 589 nm)
- Explosion Limit:0.5 - 21.8 %(V)
- Density:0.76 g/cm3 (25 °C)
- Ignition Point:340 °C
- Hazard Class:3
- Hazard Class Description:Flammable liquid