Order Entry
Germany
ContactUsLinkComponent
 
Hexamethyldisiloxane 98+%
 
undefined
Hexamethyldisiloxane 98+%
 

 

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3765 to 1.3785 (20°C, 589 nm)
  • GC:
    ≥98.0 %