Hexaethyldisiloxane
Supplier: CHEMPUR SILANE
Certificates
About this item
CAS Number:994-49-0
:
Formula:C₁₂H₃₀OSi₂
MDL Number:MFCD00026693
:246.54 g/mol
Boiling Pt:54 °C (0,1 mmHg)
Melting Pt:–115 °C
Density:0,859 g/cm³ (0 °C)
EINECS:213-619-8