1,1,3,3-Tetramethyldisiloxane
Supplier: CHEMPUR SILANE
Certificates
About this item
CAS Number:3277-26-7
:
Formula:C₄H₁₄OSi₂
MDL Number:MFCD00008256
:134.33 g/mol
Boiling Pt:71 °C (1013 hPa)
Density:0,7555 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:221-906-4