Order Entry
Germany
ContactUsLinkComponent
1,1,3,3-Tetramethyldisiloxane 97%
1,1,3,3-Tetramethyldisiloxane 97%
Certificates
undefined
1,1,3,3-Tetramethyldisiloxane 97%

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3690 to 1.3710 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

Related Information