


HMDS (1,1,1,3,3,3-hexamethyldisilazane) 98%
:999-97-3
:
:(CH₃)₃SiNHSi(CH₃)₃
:MFCD00008259
:161.39 g/mol
:13,04708
:125 °C (760 mmHg)
:–78 °C
:0.76 g/cm³
:20 °C
:Ambient
:213-668-5
:3286
:3,II
:01-2119438176-38
:GHS06
:GHS05
:GHS02
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4060 to 1.4090 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤30 APHA