Polyethylene melt index 25 g/10 min (190°C/2.16kg), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9002-88-4
:
Formula:(CH₂CH₂)n
MDL Number:MFCD00084423
Melting Pt:116 °C
Density:0.925 g/m (25 °C)
:M-Clarity™ quality level = MQ100