Order Entry
Germany
Orders LinkContactUsLinkComponent
5-Bromopyrimidine

5-Bromopyrimidine

Glass bottle for solids, 25 g
Catalog # MOLE12306755-25G
Irritant
Marketsource
Supplier:   Molekula
5-Bromopyrimidine
Catalog # MOLE12306755-25G
Supplier:  Molekula
Marketsource
CAS Number:  4595-59-9
5-Bromopyrimidine

Glass bottle for solids, 25 g

CAS Number:  4595-59-9
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:4595-59-9
  • Formula:C₄H₃BrN₂
  • MDL Number:MFCD00006117
  • Molecular Weight:158.99 g/mol
  • Boiling Pt:168 °C (727 mmHg)
  • Melting Pt:75…76 °C
  • EINECS:224-992-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.