Order Entry
Germany
ContactUsLinkComponent
Benzyl nicotinate
Benzyl nicotinate
Catalog # PROOMM0104.00
CAS Number:  
Benzyl nicotinate
Catalog # PROOMM0104.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :213.24 g/mol
Boiling Pt:179…180 °C (8 hPa)
Melting Pt:24 °C
Density:1,17 g/cm³ (20 °C)
EINECS:202-332-3
 :GHS07
CAS Number:94-44-0
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD00023584