Benzyl nicotinate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:94-44-0
:
Formula:C₁₃H₁₁NO₂
MDL Number:MFCD00023584
:213.24 g/mol
Merck Index:14,6526
Boiling Pt:179…180 °C (5 mmHg)
Melting Pt:21…23 °C
Density:1.165
Flash Pt:170 °C (338 °F)
EINECS:202-332-3
:GHS07
Storage Temperature:Ambient